Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGCTGACGAGGTGTGGGATTTA[C/T]GGGGAGGACAGCCTGTCTCTTGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604927 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTDP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CTDP1 - CTD phosphatase subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202504.1 | 1592 | Intron | NP_001189433.1 | |||
NM_001318511.1 | 1592 | Intron | NP_001305440.1 | |||
NM_004715.4 | 1592 | Intron | NP_004706.3 | |||
NM_048368.3 | 1592 | Intron | NP_430255.2 | |||
XM_011526261.1 | 1592 | Intron | XP_011524563.1 | |||
XM_017026078.1 | 1592 | Intron | XP_016881567.1 |
LOC105376875 - uncharacterized LOC105376875 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011526290.2 | 1592 | Intron | XP_011524592.1 | |||
XM_011526292.2 | 1592 | UTR 5 | XP_011524594.1 | |||
XM_017026112.1 | 1592 | Intron | XP_016881601.1 |
LOC284241 - uncharacterized LOC284241 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |