Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603336 | ||||||||||||||||||||
Literature Links: |
DNAH6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAH6 - dynein axonemal heavy chain 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001370.1 | Intron | NP_001361.1 | ||||
XM_006711956.2 | Intron | XP_006712019.1 | ||||
XM_011532649.2 | Intron | XP_011530951.1 | ||||
XM_011532650.2 | Intron | XP_011530952.1 | ||||
XM_011532652.1 | Intron | XP_011530954.1 | ||||
XM_011532653.1 | Intron | XP_011530955.1 | ||||
XM_011532654.1 | Intron | XP_011530956.1 | ||||
XM_011532655.2 | Intron | XP_011530957.1 | ||||
XM_011532657.1 | Intron | XP_011530959.1 | ||||
XM_011532660.1 | Intron | XP_011530962.1 | ||||
XM_017003521.1 | Intron | XP_016859010.1 | ||||
XM_017003522.1 | Intron | XP_016859011.1 | ||||
XM_017003523.1 | Intron | XP_016859012.1 | ||||
XM_017003524.1 | Intron | XP_016859013.1 |