Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602654 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FARP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FARP1 - FERM, ARH/RhoGEF and pleckstrin domain protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001715.3 | Intron | NP_001001715.2 | ||||
NM_001286839.1 | Intron | NP_001273768.1 | ||||
NM_005766.3 | Intron | NP_005757.1 | ||||
XM_011521046.2 | Intron | XP_011519348.1 | ||||
XM_017020312.1 | Intron | XP_016875801.1 | ||||
XM_017020313.1 | Intron | XP_016875802.1 |