Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300688 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCORL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BCORL1 - BCL6 corepressor-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184772.2 | Intron | NP_001171701.1 | ||||
NM_021946.4 | Intron | NP_068765.3 | ||||
XM_005262452.4 | Intron | XP_005262509.2 | ||||
XM_005262453.4 | Intron | XP_005262510.2 | ||||
XM_005262454.3 | Intron | XP_005262511.2 | ||||
XM_005262455.4 | Intron | XP_005262512.2 | ||||
XM_005262456.4 | Intron | XP_005262513.2 | ||||
XM_006724776.3 | Intron | XP_006724839.1 | ||||
XM_006724777.3 | Intron | XP_006724840.1 | ||||
XM_006724779.2 | Intron | XP_006724842.1 | ||||
XM_017029721.1 | Intron | XP_016885210.1 | ||||
XM_017029722.1 | Intron | XP_016885211.1 |