Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602774 MIM: 605792 | ||||||||||||||||||||
Literature Links: |
RAD51C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAD51C - RAD51 paralog C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002876.3 | Intron | NP_002867.1 | ||||
NM_058216.2 | Intron | NP_478123.1 | ||||
XM_006722001.3 | Intron | XP_006722064.1 | ||||
XM_006722002.3 | Intron | XP_006722065.1 | ||||
XM_006722004.3 | Intron | XP_006722067.1 | ||||
XM_006722005.3 | Intron | XP_006722068.1 | ||||
XM_011525092.2 | Intron | XP_011523394.1 | ||||
XM_011525093.2 | Intron | XP_011523395.1 | ||||
XM_011525094.2 | Intron | XP_011523396.1 | ||||
XM_017024914.1 | Intron | XP_016880403.1 | ||||
XM_017024915.1 | Intron | XP_016880404.1 | ||||
XM_017024916.1 | Intron | XP_016880405.1 | ||||
XM_017024917.1 | Intron | XP_016880406.1 | ||||
XM_017024918.1 | Intron | XP_016880407.1 | ||||
XM_017024919.1 | Intron | XP_016880408.1 |
TEX14 - testis expressed 14, intercellular bridge forming factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |