Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603583 MIM: 615533 | ||||||||||||||||||||
Literature Links: |
DLG2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DLG2 - discs large MAGUK scaffold protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM126B - transmembrane protein 126B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193537.2 | Intron | NP_001180466.1 | ||||
NM_001193538.2 | Intron | NP_001180467.1 | ||||
NM_001256546.1 | Intron | NP_001243475.1 | ||||
NM_001256547.1 | Intron | NP_001243476.1 | ||||
NM_018480.4 | Intron | NP_060950.3 | ||||
XM_011545165.2 | Intron | XP_011543467.1 |