Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 130592 MIM: 616408 MIM: 137020 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EEF1D PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EEF1D - eukaryotic translation elongation factor 1 delta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PYCRL - pyrroline-5-carboxylate reductase-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIGD5 - tigger transposable element derived 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSTA3 - tissue specific transplantation antigen P35B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |