Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
69 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602435 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPIE PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPIE - peptidylprolyl isomerase E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195007.1 | Intron | NP_001181936.1 | ||||
NM_001319293.1 | Intron | NP_001306222.1 | ||||
NM_006112.3 | Intron | NP_006103.1 | ||||
NM_203456.2 | Intron | NP_982281.1 | ||||
XM_006710289.2 | Intron | XP_006710352.1 | ||||
XM_006710290.3 | Intron | XP_006710353.1 | ||||
XM_011540501.1 | Intron | XP_011538803.1 | ||||
XM_017000051.1 | Intron | XP_016855540.1 | ||||
XM_017000052.1 | Intron | XP_016855541.1 |
Set Membership: |
HapMap |