Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615068 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPG5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EPG5 - ectopic P-granules autophagy protein 5 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020964.2 | Intron | NP_066015.2 | ||||
XM_011526122.1 | Intron | XP_011524424.1 | ||||
XM_017025889.1 | Intron | XP_016881378.1 | ||||
XM_017025890.1 | Intron | XP_016881379.1 | ||||
XM_017025891.1 | Intron | XP_016881380.1 | ||||
XM_017025892.1 | Intron | XP_016881381.1 | ||||
XM_017025893.1 | Intron | XP_016881382.1 |
SIGLEC15 - sialic acid binding Ig like lectin 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |