Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608592 | ||||||||||||||||||||
Literature Links: |
CTDSPL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTDSPL - CTD small phosphatase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008392.1 | 89 | Missense Mutation | AAG,ACG | K,T 14 | NP_001008393.1 | |
NM_005808.2 | 89 | Missense Mutation | AAG,ACG | K,T 14 | NP_005799.2 | |
XM_017005519.1 | 89 | Missense Mutation | AAG,ACG | K,T 14 | XP_016861008.1 | |
XM_017005520.1 | 89 | Intron | XP_016861009.1 |
ITGA9-AS1 - ITGA9 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |