Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTACAACTGCTGGGCTTGGAAGA[C/T]GGCACTGACCCCAGTGATAGGAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100996634 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100996634 - transmembrane protein FLJ37396 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277339.1 | 4167 | Intron | NP_001264268.1 | |||
XM_017010153.1 | 4167 | Silent Mutation | GAC,GAT | D,D 49 | XP_016865642.1 | |
XM_017010154.1 | 4167 | Silent Mutation | GAC,GAT | D,D 49 | XP_016865643.1 |