Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCGCGCACCACCGCCCCTTCTC[C/G]GCCCTCCTGCGTTTGCCCAGGGTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609130 MIM: 172200 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APITD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APITD1 - apoptosis-inducing, TAF9-like domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199294.2 | 381 | UTR 5 | NP_954988.1 |
APITD1-CORT - APITD1-CORT readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243768.1 | 381 | Intron | NP_001230697.1 | |||
NM_001270517.1 | 381 | UTR 5 | NP_001257446.1 | |||
NM_198544.3 | 381 | UTR 5 | NP_940946.1 | |||
NM_199006.2 | 381 | UTR 5 | NP_950171.2 |
PGD - phosphogluconate dehydrogenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |