Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf88 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C11orf88 - chromosome 11 open reading frame 88 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LAYN - layilin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258390.1 | Intron | NP_001245319.1 | ||||
NM_001258391.1 | Intron | NP_001245320.1 | ||||
NM_001318799.1 | Intron | NP_001305728.1 | ||||
NM_178834.4 | Intron | NP_849156.1 | ||||
XM_006718769.3 | Intron | XP_006718832.1 | ||||
XM_006718770.3 | Intron | XP_006718833.1 |