Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
79 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615734 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLCC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CLCC1 - chloride channel CLIC like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR47 - WD repeat domain 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142550.1 | 3689 | UTR 3 | NP_001136022.1 | |||
NM_001142551.1 | 3689 | UTR 3 | NP_001136023.1 | |||
NM_014969.5 | 3689 | UTR 3 | NP_055784.3 | |||
XM_006710460.3 | 3689 | UTR 3 | XP_006710523.1 | |||
XM_011541027.2 | 3689 | UTR 3 | XP_011539329.1 | |||
XM_011541028.2 | 3689 | UTR 3 | XP_011539330.1 | |||
XM_011541029.1 | 3689 | UTR 3 | XP_011539331.1 | |||
XM_011541030.1 | 3689 | UTR 3 | XP_011539332.1 | |||
XM_011541031.1 | 3689 | UTR 3 | XP_011539333.1 | |||
XM_017000696.1 | 3689 | UTR 3 | XP_016856185.1 | |||
XM_017000697.1 | 3689 | UTR 3 | XP_016856186.1 |