Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTTTGGGGCCAGTGCTAGGAAAT[A/G]AGTCAGAGAGAGAGGCCAAATCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLT1D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GLT1D1 - glycosyltransferase 1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144669.2 | Intron | NP_653270.1 | ||||
XM_005253553.4 | Intron | XP_005253610.2 | ||||
XM_011537956.2 | Intron | XP_011536258.2 | ||||
XM_011537957.2 | Intron | XP_011536259.1 | ||||
XM_017018855.1 | Intron | XP_016874344.1 | ||||
XM_017018856.1 | Intron | XP_016874345.1 | ||||
XM_017018857.1 | Intron | XP_016874346.1 | ||||
XM_017018858.1 | Intron | XP_016874347.1 | ||||
XM_017018859.1 | Intron | XP_016874348.1 | ||||
XM_017018860.1 | Intron | XP_016874349.1 | ||||
XM_017018861.1 | Intron | XP_016874350.1 |
LOC100128276 - uncharacterized LOC100128276 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101929984 - uncharacterized LOC101929984 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |