Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 173850 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGSF23 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGSF23 - immunoglobulin superfamily member 23 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4531 - microRNA 4531 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PVR - poliovirus receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135768.2 | 49 | UTR 5 | NP_001129240.1 | |||
NM_001135769.2 | 49 | UTR 5 | NP_001129241.1 | |||
NM_001135770.3 | 49 | UTR 5 | NP_001129242.2 | |||
NM_006505.4 | 49 | UTR 5 | NP_006496.4 |