Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600041 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
P2RY2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
P2RY2 - purinergic receptor P2Y2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002564.3 | 816 | Intron | NP_002555.3 | |||
NM_176071.2 | 816 | Intron | NP_788085.2 | |||
NM_176072.2 | 816 | Intron | NP_788086.2 | |||
XM_005274019.4 | 816 | Intron | XP_005274076.1 | |||
XM_005274020.4 | 816 | UTR 5 | XP_005274077.1 | |||
XM_005274021.4 | 816 | Intron | XP_005274078.1 | |||
XM_011545074.2 | 816 | Intron | XP_011543376.1 | |||
XM_017017839.1 | 816 | Intron | XP_016873328.1 |
Set Membership: |
HapMap |