Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGTATGTTCTAAATCCAAAAAAA[C/T]TTTAAATTTTTGTATGTTGAAAGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118190 MIM: 600141 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HSPD1 - heat shock protein family D (Hsp60) member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HSPE1 - heat shock protein family E (Hsp10) member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002157.2 | Intron | NP_002148.1 |
HSPE1-MOB4 - HSPE1-MOB4 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202485.1 | Intron | NP_001189414.1 |
Set Membership: |
HapMap |