Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604049 | ||||||||||||||||||||
Literature Links: |
XYLB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
XYLB - xylulokinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005108.3 | Intron | NP_005099.2 | ||||
XM_011534325.2 | Intron | XP_011532627.1 | ||||
XM_011534327.1 | Intron | XP_011532629.1 | ||||
XM_011534328.2 | Intron | XP_011532630.1 | ||||
XM_011534329.1 | Intron | XP_011532631.1 | ||||
XM_011534330.2 | Intron | XP_011532632.1 | ||||
XM_017007595.1 | Intron | XP_016863084.1 | ||||
XM_017007596.1 | Intron | XP_016863085.1 | ||||
XM_017007597.1 | Intron | XP_016863086.1 | ||||
XM_017007598.1 | Intron | XP_016863087.1 | ||||
XM_017007599.1 | Intron | XP_016863088.1 | ||||
XM_017007600.1 | Intron | XP_016863089.1 |