Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612477 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IQCG PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IQCG - IQ motif containing G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134435.2 | Intron | NP_001127907.1 | ||||
NM_001323027.1 | Intron | NP_001309956.1 | ||||
NM_001323028.1 | Intron | NP_001309957.1 | ||||
NM_001323029.1 | Intron | NP_001309958.1 | ||||
NM_001323030.1 | Intron | NP_001309959.1 | ||||
NM_032263.4 | Intron | NP_115639.1 | ||||
XM_017007316.1 | Intron | XP_016862805.1 | ||||
XM_017007317.1 | Intron | XP_016862806.1 |
LRCH3 - leucine rich repeats and calponin homology domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |