Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
77 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602923 MIM: 611750 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBBP4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RBBP4 - RB binding protein 4, chromatin remodeling factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135255.1 | 3174 | UTR 3 | NP_001128727.1 | |||
NM_001135256.1 | 3174 | UTR 3 | NP_001128728.1 | |||
NM_005610.2 | 3174 | UTR 3 | NP_005601.1 |
SYNC - syncoilin, intermediate filament protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161708.1 | 3174 | UTR 3 | NP_001155180.1 | |||
NM_030786.2 | 3174 | UTR 3 | NP_110413.2 |