Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM87A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TMEM87A - transmembrane protein 87A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110503.2 | Intron | NP_001103973.1 | ||||
NM_001286487.1 | Intron | NP_001273416.1 | ||||
NM_015497.4 | Intron | NP_056312.2 | ||||
XM_005254287.1 | Intron | XP_005254344.1 | ||||
XM_006720468.2 | Intron | XP_006720531.1 | ||||
XM_011521443.2 | Intron | XP_011519745.1 | ||||
XM_011521444.2 | Intron | XP_011519746.1 | ||||
XM_017022067.1 | Intron | XP_016877556.1 |