Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612188 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM87A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM87A - transmembrane protein 87A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110503.2 | 2718 | Intron | NP_001103973.1 | |||
NM_001286487.1 | 2718 | UTR 3 | NP_001273416.1 | |||
NM_015497.4 | 2718 | UTR 3 | NP_056312.2 | |||
XM_005254287.1 | 2718 | UTR 3 | XP_005254344.1 | |||
XM_006720468.2 | 2718 | Intron | XP_006720531.1 | |||
XM_011521443.2 | 2718 | Intron | XP_011519745.1 | |||
XM_011521444.2 | 2718 | Intron | XP_011519746.1 | |||
XM_017022067.1 | 2718 | Intron | XP_016877556.1 |
VPS39 - VPS39, HOPS complex subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |