Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610795 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SORBS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SORBS3 - sorbin and SH3 domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018003.2 | Intron | NP_001018003.1 | ||||
NM_005775.4 | Intron | NP_005766.3 | ||||
XM_005273371.1 | Intron | XP_005273428.1 | ||||
XM_006716266.1 | Intron | XP_006716329.1 | ||||
XM_006716268.2 | Intron | XP_006716331.1 | ||||
XM_017012944.1 | Intron | XP_016868433.1 | ||||
XM_017012945.1 | Intron | XP_016868434.1 | ||||
XM_017012946.1 | Intron | XP_016868435.1 |