Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604054 MIM: 602170 | ||||||||||||||||||||
Literature Links: |
ACAA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACAA1 - acetyl-CoA acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYD88 - myeloid differentiation primary response 88 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172566.1 | 145 | UTR 5 | NP_001166037.1 | |||
NM_001172567.1 | 145 | UTR 5 | NP_001166038.1 | |||
NM_001172568.1 | 145 | UTR 5 | NP_001166039.1 | |||
NM_001172569.1 | 145 | UTR 5 | NP_001166040.1 | |||
NM_002468.4 | 145 | UTR 5 | NP_002459.2 | |||
XM_005265172.1 | 145 | UTR 5 | XP_005265229.1 | |||
XM_006713170.1 | 145 | Intron | XP_006713233.1 |