Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616102 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC102B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCDC102B - coiled-coil domain containing 102B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001093729.1 | Intron | NP_001087198.1 | ||||
NM_024781.2 | Intron | NP_079057.2 | ||||
XM_011526174.2 | Intron | XP_011524476.1 | ||||
XM_011526175.2 | Intron | XP_011524477.1 | ||||
XM_017025973.1 | Intron | XP_016881462.1 | ||||
XM_017025974.1 | Intron | XP_016881463.1 | ||||
XM_017025975.1 | Intron | XP_016881464.1 | ||||
XM_017025976.1 | Intron | XP_016881465.1 | ||||
XM_017025977.1 | Intron | XP_016881466.1 | ||||
XM_017025978.1 | Intron | XP_016881467.1 | ||||
XM_017025979.1 | Intron | XP_016881468.1 | ||||
XM_017025980.1 | Intron | XP_016881469.1 |
TMX3 - thioredoxin related transmembrane protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019022.3 | Intron | NP_061895.3 | ||||
XM_005266714.2 | Intron | XP_005266771.1 | ||||
XM_011526033.2 | Intron | XP_011524335.1 | ||||
XM_011526034.2 | Intron | XP_011524336.1 | ||||
XM_011526035.2 | Intron | XP_011524337.1 |
Set Membership: |
HapMap |