Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTGACTAGATTAATGTGAGACTT[A/C]ACATTTGACTTGATTCATGGTAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606168 | ||||||||||||||||||||
Literature Links: |
DDX20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDX20 - DEAD-box helicase 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007204.4 | Intron | NP_009135.4 |
FAM212B - family with sequence similarity 212 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM212B-AS1 - FAM212B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101928718 - uncharacterized LOC101928718 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |