Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611089 | ||||||||||||||||||||
Literature Links: |
MTMR14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTMR14 - myotubularin related protein 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077525.2 | Intron | NP_001070993.1 | ||||
NM_001077526.2 | Intron | NP_001070994.1 | ||||
NM_022485.4 | Intron | NP_071930.2 | ||||
XM_005265400.1 | Intron | XP_005265457.1 | ||||
XM_011534012.1 | Intron | XP_011532314.1 | ||||
XM_011534013.1 | Intron | XP_011532315.1 | ||||
XM_017007034.1 | Intron | XP_016862523.1 | ||||
XM_017007035.1 | Intron | XP_016862524.1 | ||||
XM_017007036.1 | Intron | XP_016862525.1 | ||||
XM_017007037.1 | Intron | XP_016862526.1 | ||||
XM_017007038.1 | Intron | XP_016862527.1 | ||||
XM_017007039.1 | Intron | XP_016862528.1 | ||||
XM_017007040.1 | Intron | XP_016862529.1 | ||||
XM_017007041.1 | Intron | XP_016862530.1 | ||||
XM_017007042.1 | Intron | XP_016862531.1 | ||||
XM_017007043.1 | Intron | XP_016862532.1 | ||||
XM_017007044.1 | Intron | XP_016862533.1 | ||||
XM_017007045.1 | Intron | XP_016862534.1 |