Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614348 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADTRP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ADTRP - androgen dependent TFPI regulating protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143948.1 | Intron | NP_001137420.1 | ||||
NM_032744.3 | Intron | NP_116133.1 | ||||
XM_005249454.3 | Intron | XP_005249511.1 | ||||
XM_011514956.1 | Intron | XP_011513258.1 | ||||
XM_011514958.2 | Intron | XP_011513260.1 | ||||
XM_011514959.1 | Intron | XP_011513261.1 | ||||
XM_017011377.1 | Intron | XP_016866866.1 |
LOC340184 - uncharacterized LOC340184 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |