Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
58 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605037 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF17 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIF17 - kinesin family member 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH2D5 - SH2 domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001103160.1 | 2545 | UTR 3 | NP_001096630.1 | |||
NM_001103161.1 | 2545 | UTR 3 | NP_001096631.1 | |||
XM_011541459.1 | 2545 | UTR 3 | XP_011539761.1 | |||
XM_011541460.1 | 2545 | UTR 3 | XP_011539762.1 | |||
XM_011541461.2 | 2545 | UTR 3 | XP_011539763.1 | |||
XM_011541462.1 | 2545 | UTR 3 | XP_011539764.1 |
Set Membership: |
HapMap |