Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602923 MIM: 615891 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBBP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RBBP4 - RB binding protein 4, chromatin remodeling factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135255.1 | Intron | NP_001128727.1 | ||||
NM_001135256.1 | Intron | NP_001128728.1 | ||||
NM_005610.2 | Intron | NP_005601.1 |
ZBTB8OS - zinc finger and BTB domain containing 8 opposite strand | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308135.1 | Intron | NP_001295064.1 | ||||
NM_001308136.1 | Intron | NP_001295065.1 | ||||
NM_001308137.1 | Intron | NP_001295066.1 | ||||
NM_001308138.1 | Intron | NP_001295067.1 | ||||
NM_001308139.1 | Intron | NP_001295068.1 | ||||
NM_001308140.1 | Intron | NP_001295069.1 | ||||
NM_001308141.1 | Intron | NP_001295070.1 | ||||
NM_178547.3 | Intron | NP_848642.1 | ||||
XM_011541327.2 | Intron | XP_011539629.1 | ||||
XM_011541328.1 | Intron | XP_011539630.1 | ||||
XM_011541329.2 | Intron | XP_011539631.1 | ||||
XM_011541330.2 | Intron | XP_011539632.1 | ||||
XM_011541334.2 | Intron | XP_011539636.1 | ||||
XM_011541335.2 | Intron | XP_011539637.1 | ||||
XM_011541337.2 | Intron | XP_011539639.1 | ||||
XM_011541338.2 | Intron | XP_011539640.1 | ||||
XM_017001134.1 | Intron | XP_016856623.1 | ||||
XM_017001135.1 | Intron | XP_016856624.1 | ||||
XM_017001136.1 | Intron | XP_016856625.1 | ||||
XM_017001137.1 | Intron | XP_016856626.1 | ||||
XM_017001138.1 | Intron | XP_016856627.1 |