Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
73 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600172 MIM: 612276 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf122 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C1orf122 - chromosome 1 open reading frame 122 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142726.1 | 7573 | Intron | NP_001136198.1 | |||
NM_198446.2 | 7573 | Intron | NP_940848.2 |
MANEAL - mannosidase endo-alpha like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTF1 - metal regulatory transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005955.2 | 7573 | UTR 3 | NP_005946.2 | |||
XM_011541491.2 | 7573 | UTR 3 | XP_011539793.1 | |||
XM_011541493.2 | 7573 | Intron | XP_011539795.1 | |||
XM_011541494.1 | 7573 | UTR 3 | XP_011539796.1 |
YRDC - yrdC N6-threonylcarbamoyltransferase domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |