Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
50 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123691 MIM: 611245 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRYZ PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CRYZ - crystallin zeta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TYW3 - tRNA-yW synthesizing protein 3 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162916.1 | Intron | NP_001156388.1 | ||||
NM_138467.2 | Intron | NP_612476.1 | ||||
XM_006710347.1 | Intron | XP_006710410.1 |
Set Membership: |
HapMap |