Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605439 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EHF PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
EHF - ETS homologous factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206615.1 | Intron | NP_001193544.1 | ||||
NM_001206616.1 | Intron | NP_001193545.1 | ||||
NM_012153.5 | Intron | NP_036285.2 | ||||
XM_005252857.1 | Intron | XP_005252914.1 | ||||
XM_005252860.1 | Intron | XP_005252917.1 | ||||
XM_005252861.1 | Intron | XP_005252918.1 | ||||
XM_005252862.1 | Intron | XP_005252919.1 | ||||
XM_011519983.1 | Intron | XP_011518285.1 | ||||
XM_011519984.1 | Intron | XP_011518286.1 | ||||
XM_011519985.1 | Intron | XP_011518287.1 | ||||
XM_017017534.1 | Intron | XP_016873023.1 |
Set Membership: |
HapMap Validated |