Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGGATGGCAAATGTGCACAAGTG[C/T]CAATTCCTATAAAACTGGCTCGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151670 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LIPC - lipase C, hepatic type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000236.2 | Intron | NP_000227.2 | ||||
XM_005254372.1 | Intron | XP_005254429.1 | ||||
XM_005254374.4 | Intron | XP_005254431.2 | ||||
XM_006720502.3 | Intron | XP_006720565.1 | ||||
XM_017022176.1 | Intron | XP_016877665.1 |
LOC101928635 - uncharacterized LOC101928635 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011522293.2 | Intron | XP_011520595.1 |
Set Membership: |
HapMap |