Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600126 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE4A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
PDE4A - phosphodiesterase 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111307.1 | Intron | NP_001104777.1 | ||||
NM_001111308.1 | Intron | NP_001104778.1 | ||||
NM_001111309.1 | Intron | NP_001104779.1 | ||||
NM_001243121.1 | Intron | NP_001230050.1 | ||||
NM_006202.2 | Intron | NP_006193.1 | ||||
XM_005259933.2 | Intron | XP_005259990.1 | ||||
XM_011528054.1 | Intron | XP_011526356.1 | ||||
XM_011528055.2 | Intron | XP_011526357.1 | ||||
XM_017026865.1 | Intron | XP_016882354.1 |
Set Membership: |
HapMap Validated |