Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603261 MIM: 602176 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIP4K2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PIP4K2B - phosphatidylinositol-5-phosphate 4-kinase type 2 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003559.4 | Intron | NP_003550.1 | ||||
XM_011525326.2 | Intron | XP_011523628.1 | ||||
XM_011525327.2 | Intron | XP_011523629.1 | ||||
XM_011525329.2 | Intron | XP_011523631.1 | ||||
XM_011525330.2 | Intron | XP_011523632.1 | ||||
XM_017025196.1 | Intron | XP_016880685.1 | ||||
XM_017025197.1 | Intron | XP_016880686.1 | ||||
XM_017025198.1 | Intron | XP_016880687.1 | ||||
XM_017025199.1 | Intron | XP_016880688.1 |
PSMB3 - proteasome subunit beta 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |