Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114350 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIF1L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AIF1L - allograft inflammatory factor 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUP214 - nucleoporin 214 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318324.1 | Intron | NP_001305253.1 | ||||
NM_001318325.1 | Intron | NP_001305254.1 | ||||
NM_005085.3 | Intron | NP_005076.3 | ||||
XM_005272216.3 | Intron | XP_005272273.1 | ||||
XM_005272218.3 | Intron | XP_005272275.1 | ||||
XM_005272219.3 | Intron | XP_005272276.1 | ||||
XM_006717292.3 | Intron | XP_006717355.1 | ||||
XM_011519053.2 | Intron | XP_011517355.1 | ||||
XM_011519055.2 | Intron | XP_011517357.1 | ||||
XM_017015167.1 | Intron | XP_016870656.1 |
Set Membership: |
HapMap |