Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGCTTTTTGGTTATCTGATTATT[A/C]TTCTTGATTTCCTTCATGATCCTCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CWF19L2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CWF19L2 - CWF19-like 2, cell cycle control (S. pombe) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152434.2 | Intron | NP_689647.2 | ||||
XM_011542620.2 | Intron | XP_011540922.1 | ||||
XM_017017239.1 | Intron | XP_016872728.1 |
LOC105369477 - uncharacterized LOC105369477 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |