Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMC8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARMC8 - armadillo repeat containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267041.2 | 53 | UTR 5 | NP_001253970.1 | |||
NM_001267042.2 | 53 | UTR 5 | NP_001253971.1 | |||
NM_001282342.1 | 53 | UTR 5 | NP_001269271.1 | |||
NM_014154.4 | 53 | UTR 5 | NP_054873.2 | |||
NM_015396.5 | 53 | UTR 5 | NP_056211.2 | |||
NM_213654.3 | 53 | UTR 5 | NP_998819.1 | |||
XM_006713564.3 | 53 | UTR 5 | XP_006713627.1 | |||
XM_006713565.2 | 53 | Intron | XP_006713628.1 | |||
XM_006713566.3 | 53 | UTR 5 | XP_006713629.1 | |||
XM_006713567.3 | 53 | UTR 5 | XP_006713630.1 | |||
XM_011512632.1 | 53 | UTR 5 | XP_011510934.1 | |||
XM_011512633.1 | 53 | UTR 5 | XP_011510935.1 | |||
XM_017006095.1 | 53 | Intron | XP_016861584.1 | |||
XM_017006096.1 | 53 | UTR 5 | XP_016861585.1 | |||
XM_017006097.1 | 53 | UTR 5 | XP_016861586.1 | |||
XM_017006098.1 | 53 | UTR 5 | XP_016861587.1 | |||
XM_017006099.1 | 53 | Intron | XP_016861588.1 | |||
XM_017006100.1 | 53 | UTR 5 | XP_016861589.1 | |||
XM_017006101.1 | 53 | Intron | XP_016861590.1 |