Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613311 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYRM4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LYRM4 - LYR motif containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164840.2 | Intron | NP_001158312.1 | ||||
NM_001164841.2 | Intron | NP_001158313.1 | ||||
NM_001318782.1 | Intron | NP_001305711.1 | ||||
NM_001318783.1 | Intron | NP_001305712.1 | ||||
NM_020408.5 | Intron | NP_065141.3 | ||||
XM_017011083.1 | Intron | XP_016866572.1 | ||||
XM_017011084.1 | Intron | XP_016866573.1 |
LYRM4-AS1 - LYRM4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |