Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604811 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LILRB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LILRB1 - leukocyte immunoglobulin like receptor B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081637.2 | Intron | NP_001075106.2 | ||||
NM_001081638.3 | Intron | NP_001075107.2 | ||||
NM_001081639.3 | Intron | NP_001075108.2 | ||||
NM_001278398.2 | Intron | NP_001265327.2 | ||||
NM_001278399.2 | Intron | NP_001265328.2 | ||||
NM_006669.6 | Intron | NP_006660.4 | ||||
XM_011526331.2 | Intron | XP_011524633.1 | ||||
XM_011526332.2 | Intron | XP_011524634.1 | ||||
XM_011526335.2 | Intron | XP_011524637.1 | ||||
XM_011526336.2 | Intron | XP_011524638.1 | ||||
XM_011526338.2 | Intron | XP_011524640.1 | ||||
XM_017026182.1 | Intron | XP_016881671.1 | ||||
XM_017026183.1 | Intron | XP_016881672.1 | ||||
XM_017026184.1 | Intron | XP_016881673.1 | ||||
XM_017026185.1 | Intron | XP_016881674.1 | ||||
XM_017026186.1 | Intron | XP_016881675.1 | ||||
XM_017026187.1 | Intron | XP_016881676.1 | ||||
XM_017026188.1 | Intron | XP_016881677.1 | ||||
XM_017026189.1 | Intron | XP_016881678.1 | ||||
XM_017026190.1 | Intron | XP_016881679.1 | ||||
XM_017026191.1 | Intron | XP_016881680.1 | ||||
XM_017026192.1 | Intron | XP_016881681.1 |
Set Membership: |
HapMap Validated |