Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
49 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613360 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRAM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DRAM2 - DNA damage regulated autophagy modulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178454.4 | 2024 | UTR 3 | NP_848549.3 | |||
XM_005270469.2 | 2024 | UTR 3 | XP_005270526.1 | |||
XM_005270470.1 | 2024 | UTR 3 | XP_005270527.1 | |||
XM_006710361.1 | 2024 | UTR 3 | XP_006710424.1 | |||
XM_006710362.1 | 2024 | UTR 3 | XP_006710425.1 | |||
XM_011540707.1 | 2024 | UTR 3 | XP_011539009.1 | |||
XM_011540708.1 | 2024 | UTR 3 | XP_011539010.1 | |||
XM_017000320.1 | 2024 | UTR 3 | XP_016855809.1 | |||
XM_017000321.1 | 2024 | UTR 3 | XP_016855810.1 |