Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF329 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF329 - zinc finger protein 329 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024620.3 | 1299 | Silent Mutation | GAC,GAT | D,D 354 | NP_078896.3 | |
XM_006723381.2 | 1299 | Silent Mutation | GAC,GAT | D,D 354 | XP_006723444.1 | |
XM_006723382.3 | 1299 | Silent Mutation | GAC,GAT | D,D 354 | XP_006723445.1 | |
XM_006723383.3 | 1299 | Silent Mutation | GAC,GAT | D,D 354 | XP_006723446.1 | |
XM_006723384.3 | 1299 | Silent Mutation | GAC,GAT | D,D 354 | XP_006723447.1 | |
XM_011527310.2 | 1299 | Silent Mutation | GAC,GAT | D,D 377 | XP_011525612.1 | |
XM_011527311.2 | 1299 | Silent Mutation | GAC,GAT | D,D 377 | XP_011525613.1 | |
XM_011527312.2 | 1299 | Silent Mutation | GAC,GAT | D,D 377 | XP_011525614.1 | |
XM_011527313.2 | 1299 | Silent Mutation | GAC,GAT | D,D 377 | XP_011525615.1 | |
XM_011527314.2 | 1299 | Silent Mutation | GAC,GAT | D,D 377 | XP_011525616.1 | |
XM_011527315.1 | 1299 | Silent Mutation | GAC,GAT | D,D 354 | XP_011525617.1 | |
XM_011527316.2 | 1299 | Silent Mutation | GAC,GAT | D,D 354 | XP_011525618.1 | |
XM_017027307.1 | 1299 | Silent Mutation | GAC,GAT | D,D 354 | XP_016882796.1 | |
XM_017027308.1 | 1299 | Silent Mutation | GAC,GAT | D,D 354 | XP_016882797.1 | |
XM_017027309.1 | 1299 | Silent Mutation | GAC,GAT | D,D 354 | XP_016882798.1 | |
XM_017027310.1 | 1299 | Silent Mutation | GAC,GAT | D,D 354 | XP_016882799.1 | |
XM_017027311.1 | 1299 | Silent Mutation | GAC,GAT | D,D 354 | XP_016882800.1 |
Set Membership: |
HapMap |