Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605117 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SOCS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SOCS2 - suppressor of cytokine signaling 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270467.1 | Intron | NP_001257396.1 | ||||
NM_001270468.1 | Intron | NP_001257397.1 | ||||
NM_001270469.1 | Intron | NP_001257398.1 | ||||
NM_001270470.1 | Intron | NP_001257399.1 | ||||
NM_001270471.1 | Intron | NP_001257400.1 | ||||
NM_003877.4 | Intron | NP_003868.1 | ||||
XM_011538929.1 | Intron | XP_011537231.1 | ||||
XM_011538935.1 | Intron | XP_011537237.1 | ||||
XM_011538936.1 | Intron | XP_011537238.1 | ||||
XM_017020147.1 | Intron | XP_016875636.1 | ||||
XM_017020148.1 | Intron | XP_016875637.1 | ||||
XM_017020149.1 | Intron | XP_016875638.1 | ||||
XM_017020150.1 | Intron | XP_016875639.1 | ||||
XM_017020151.1 | Intron | XP_016875640.1 | ||||
XM_017020152.1 | Intron | XP_016875641.1 | ||||
XM_017020153.1 | Intron | XP_016875642.1 | ||||
XM_017020154.1 | Intron | XP_016875643.1 | ||||
XM_017020155.1 | Intron | XP_016875644.1 | ||||
XM_017020156.1 | Intron | XP_016875645.1 |
SOCS2-AS1 - SOCS2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |