Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609866 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STARD13 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
STARD13 - StAR related lipid transfer domain containing 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243466.1 | 5825 | Intron | NP_001230395.1 | |||
NM_001243474.1 | 5825 | Intron | NP_001230403.1 | |||
NM_001243476.2 | 5825 | UTR 3 | NP_001230405.1 | |||
NM_052851.2 | 5825 | UTR 3 | NP_443083.1 | |||
NM_178006.3 | 5825 | UTR 3 | NP_821074.1 | |||
NM_178007.2 | 5825 | UTR 3 | NP_821075.1 | |||
XM_011535299.2 | 5825 | UTR 3 | XP_011533601.1 | |||
XM_017020834.1 | 5825 | UTR 3 | XP_016876323.1 | |||
XM_017020835.1 | 5825 | UTR 3 | XP_016876324.1 |
Set Membership: |
HapMap |