Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGCAGACCTGCACCCACCCTCA[A/G]GATACCTCCCAGGACAGGCTACTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604368 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPNMB PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
GPNMB - glycoprotein nmb | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005340.1 | Intron | NP_001005340.1 | ||||
NM_002510.2 | Intron | NP_002501.1 | ||||
XM_005249578.2 | Intron | XP_005249635.1 | ||||
XM_017011676.1 | Intron | XP_016867165.1 | ||||
XM_017011677.1 | Intron | XP_016867166.1 | ||||
XM_017011678.1 | Intron | XP_016867167.1 |
LOC101927890 - uncharacterized LOC101927890 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |