Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612680 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CELF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CELF5 - CUGBP, Elav-like family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172673.1 | Intron | NP_001166144.1 | ||||
NM_021938.3 | Intron | NP_068757.2 | ||||
XM_006722832.1 | Intron | XP_006722895.1 | ||||
XM_006722833.1 | Intron | XP_006722896.1 | ||||
XM_006722836.2 | Intron | XP_006722899.1 | ||||
XM_006722837.3 | Intron | XP_006722900.1 | ||||
XM_006722838.3 | Intron | XP_006722901.1 | ||||
XM_011528173.1 | Intron | XP_011526475.1 | ||||
XM_011528174.2 | Intron | XP_011526476.1 | ||||
XM_017027111.1 | Intron | XP_016882600.1 |