Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612135 MIM: 606730 | ||||||||||||||||||||
Literature Links: |
CABYR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CABYR - calcium binding tyrosine phosphorylation regulated | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OSBPL1A - oxysterol binding protein like 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242508.1 | Intron | NP_001229437.1 | ||||
NM_018030.4 | Intron | NP_060500.3 | ||||
NM_080597.3 | Intron | NP_542164.2 | ||||
XM_006722380.3 | Intron | XP_006722443.1 | ||||
XM_006722382.3 | Intron | XP_006722445.1 | ||||
XM_017025530.1 | Intron | XP_016881019.1 | ||||
XM_017025531.1 | Intron | XP_016881020.1 | ||||
XM_017025532.1 | Intron | XP_016881021.1 | ||||
XM_017025533.1 | Intron | XP_016881022.1 | ||||
XM_017025534.1 | Intron | XP_016881023.1 |