Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610745 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC33 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC33 - coiled-coil domain containing 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287181.1 | Intron | NP_001274110.1 | ||||
NM_025055.4 | Intron | NP_079331.3 | ||||
NM_182791.3 | Intron | NP_877592.2 | ||||
XM_005254692.1 | Intron | XP_005254749.1 | ||||
XM_011522085.2 | Intron | XP_011520387.1 | ||||
XM_011522086.1 | Intron | XP_011520388.1 | ||||
XM_011522087.2 | Intron | XP_011520389.1 | ||||
XM_011522088.1 | Intron | XP_011520390.1 | ||||
XM_011522089.2 | Intron | XP_011520391.1 | ||||
XM_011522090.2 | Intron | XP_011520392.2 | ||||
XM_017022623.1 | Intron | XP_016878112.1 | ||||
XM_017022624.1 | Intron | XP_016878113.1 | ||||
XM_017022625.1 | Intron | XP_016878114.1 | ||||
XM_017022626.1 | Intron | XP_016878115.1 | ||||
XM_017022627.1 | Intron | XP_016878116.1 | ||||
XM_017022628.1 | Intron | XP_016878117.1 | ||||
XM_017022629.1 | Intron | XP_016878118.1 | ||||
XM_017022630.1 | Intron | XP_016878119.1 | ||||
XM_017022631.1 | Intron | XP_016878120.1 | ||||
XM_017022632.1 | Intron | XP_016878121.1 | ||||
XM_017022633.1 | Intron | XP_016878122.1 |
STRA6 - stimulated by retinoic acid 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142617.1 | Intron | NP_001136089.1 | ||||
NM_001142618.1 | Intron | NP_001136090.1 | ||||
NM_001142619.1 | Intron | NP_001136091.1 | ||||
NM_001142620.1 | Intron | NP_001136092.1 | ||||
NM_001199040.1 | Intron | NP_001185969.1 | ||||
NM_001199041.1 | Intron | NP_001185970.1 | ||||
NM_001199042.1 | Intron | NP_001185971.1 | ||||
NM_022369.3 | Intron | NP_071764.3 | ||||
XM_011521883.1 | Intron | XP_011520185.1 | ||||
XM_011521884.1 | Intron | XP_011520186.1 | ||||
XM_011521885.2 | Intron | XP_011520187.1 | ||||
XM_017022478.1 | Intron | XP_016877967.1 | ||||
XM_017022479.1 | Intron | XP_016877968.1 | ||||
XM_017022480.1 | Intron | XP_016877969.1 |